Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142701.1 | NP_001136173.1 | 15 - 16 | 2058 | 78 | |
NM_027521.3 | NP_081797.1 | 14 - 15 | 1610 | 78 | ||
GenBank mRNA | AK018130.1 | - | 2 - 3 | 125 | 78 | |
AK044860.1 | - | 14 - 15 | 1572 | 78 | ||
AK134029.1 | - | 8 - 9 | 1032 | 78 | ||
AK151423.1 | - | 15 - 16 | 2002 | 78 | ||
AK159194.1 | - | 15 - 16 | 2059 | 78 | ||
AK159474.1 | - | 14 - 15 | 2103 | 78 | ||
AK161766.1 | - | 7 - 8 | 1118 | 78 | ||
AK170262.1 | - | 15 - 16 | 2044 | 78 | ||
AK171304.1 | - | 15 - 16 | 2044 | 78 | ||
BC053750.1 | - | 14 - 15 | 1610 | 6333971 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |