Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142701.1 | NP_001136173.1 | 17 - 18 | 2339 | 71 | |
NM_027521.3 | NP_081797.1 | 16 - 17 | 1891 | 71 | ||
GenBank mRNA | AK018130.1 | - | 4 - 5 | 406 | 71 | |
AK044860.1 | - | 16 - 17 | 1853 | 71 | ||
AK134029.1 | - | 10 - 11 | 1313 | 71 | ||
AK151423.1 | - | 17 - 18 | 2283 | 71 | ||
AK159194.1 | - | 17 - 18 | 2340 | 71 | ||
AK159474.1 | - | 16 - 17 | 2384 | 71 | ||
AK161766.1 | - | 9 - 10 | 1399 | 71 | ||
AK170262.1 | - | 17 - 18 | 2325 | 71 | ||
AK171304.1 | - | 17 - 18 | 2325 | 71 | ||
BC053750.1 | - | 16 - 17 | 1891 | 6333971 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |