Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142701.1 | NP_001136173.1 | 18 - 19 | 2524 | 104 | |
NM_027521.3 | NP_081797.1 | 17 - 18 | 2076 | 104 | ||
GenBank mRNA | AK018130.1 | - | 5 - 6 | 591 | 104 | |
AK044860.1 | - | 17 - 18 | 2038 | 104 | ||
AK134029.1 | - | 11 - 12 | 1498 | 104 | ||
AK151423.1 | - | 18 - 19 | 2468 | 104 | ||
AK159194.1 | - | 18 - 19 | 2525 | 104 | ||
AK159474.1 | - | 17 - 18 | 2569 | 104 | ||
AK161766.1 | - | 10 - 11 | 1584 | 104 | ||
AK170262.1 | - | 18 - 19 | 2510 | 104 | ||
AK171304.1 | - | 18 - 19 | 2510 | 104 | ||
BC053750.1 | - | 17 - 18 | 2076 | 6333971 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |