Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198645.2 | NP_941047.1 | 1 - 2 | 476 | 114 | |
GenBank mRNA | AK135450.1 | - | 1 - 2 | 56 | 114 | |
BC024406.1 | - | 1 - 2 | 40 | 114 | ||
BC024862.1 | - | 1 - 2 | 451 | 5356370 | 114 | |
CB949450.1 | - | 1 - 2 | 479 | 114 | ||
CF617453.1 | - | 1 - 2 | 454 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |