Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109750.1 | NP_001103220.1 | 7 - 8 | 1256 | 75 | |
NM_010048.3 | NP_034178.2 | 7 - 8 | 1265 | 75 | ||
GenBank mRNA | AK129070.1 | - | 8 - 9 | 1115 | 75 | |
AK133880.1 | - | 4 - 5 | 554 | 75 | ||
AK141594.1 | - | 7 - 8 | 1231 | 75 | ||
AK145495.1 | - | 7 - 8 | 1182 | 75 | ||
AK153026.1 | - | 7 - 8 | 1052 | 75 | ||
AK161022.1 | - | 2016 | 75 | |||
AK161359.1 | - | 7 - 8 | 981 | 75 | ||
AK163872.1 | - | 7 - 8 | 75 | |||
AK163903.1 | - | 7 - 8 | 1194 | 75 | ||
BC060636.1 | - | 7 - 8 | 1206 | 6849298 | 75 | |
BC062978.1 | - | 7 - 8 | 1160 | 30356000 | 75 | |
D78641.1 | - | 7 - 8 | 1190 | 75 | ||
X95480.1 | - | 7 - 8 | 1131 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |