Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033324.2 | NP_201581.2 | 11 - 12 | 2340 | 89 | |
GenBank mRNA | AB086857.1 | - | 11 - 12 | 2340 | 89 | |
AK086233.1 | - | 10 - 11 | 2532 | 89 | ||
AK161410.1 | - | 3 - 4 | 338 | 89 | ||
BC051013.1 | - | 9 - 10 | 2526 | 89 | ||
BC062919.1 | - | 11 - 12 | 2360 | 6833280 | 89 | |
BC066118.1 | - | 11 - 12 | 2387 | 6400561 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |