Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033324.2 | NP_201581.2 | 13 - 14 | 2582 | 92 | |
GenBank mRNA | AB086857.1 | - | 13 - 14 | 2582 | 92 | |
AK086233.1 | - | 12 - 13 | 2774 | 92 | ||
AK161410.1 | - | 5 - 6 | 580 | 92 | ||
BC051013.1 | - | 11 - 12 | 2768 | 92 | ||
BC062919.1 | - | 13 - 14 | 2602 | 6833280 | 92 | |
BC066118.1 | - | 13 - 14 | 2629 | 6400561 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |