Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033324.2 | NP_201581.2 | 8 - 9 | 2053 | 101 | |
GenBank mRNA | AB086857.1 | - | 8 - 9 | 2053 | 101 | |
AK048056.1 | - | 7 - 8 | 1854 | 101 | ||
AK086233.1 | - | 7 - 8 | 2245 | 101 | ||
BC051013.1 | - | 6 - 7 | 2239 | 101 | ||
BC062919.1 | - | 8 - 9 | 2073 | 6833280 | 101 | |
BC066118.1 | - | 8 - 9 | 2100 | 6400561 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |