Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dis3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001295.2 | NP_001001295.2 | 10 - 11 | 1705 | 87 | |
NM_001177784.1 | NP_001171255.1 | 10 - 11 | 1666 | 87 | ||
NM_172519.3 | NP_766107.1 | 10 - 11 | 1609 | 87 | ||
GenBank mRNA | AK029974.1 | - | 10 - 11 | 1610 | 87 | |
AK032451.1 | - | 10 - 11 | 1614 | 87 | ||
AK077031.1 | - | 10 - 11 | 1669 | 87 | ||
AK150851.1 | - | 10 - 11 | 1655 | 87 | ||
AK151780.1 | - | 10 - 11 | 1655 | 87 | ||
AK153433.1 | - | 10 - 11 | 1655 | 87 | ||
AK173310.1 | - | 10 - 11 | 1639 | 87 | ||
BC056939.1 | - | 10 - 11 | 1638 | 5708685 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |