Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034895.3 | NP_001030067.2 | 10 - 11 | 1584 | 126 | |
GenBank mRNA | AK122438.1 | - | 3 - 4 | 3192 | 126 | |
BC072571.1 | - | 9 - 10 | 2253 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |