Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034895.3 | NP_001030067.2 | 8 - 9 | 1309 | 81 | |
GenBank mRNA | AK033649.1 | - | 1 - 2 | 58 | 81 | |
BC072571.1 | - | 7 - 8 | 1978 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |