Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam81a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029784.2 | NP_084060.1 | 5 - 6 | 865 | 74 | |
GenBank mRNA | AK038532.1 | - | 5 - 6 | 824 | 74 | |
AK044090.1 | - | 5 - 6 | 865 | 74 | ||
AK046408.1 | - | 5 - 6 | 758 | 74 | ||
AK079541.1 | - | 5 - 6 | 825 | 74 | ||
AK135091.1 | - | 4 - 5 | 580 | 74 | ||
BC025646.1 | - | 5 - 6 | 891 | 5324986 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |