Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026665.4 | NP_080941.3 | 3 - 4 | 592 | 150 | |
NR_037622.1 | 3 - 4 | 852 | 150 | |||
GenBank mRNA | AK016484.1 | - | 3 - 4 | 376 | 150 | |
AK019414.1 | - | 3 - 4 | 472 | 150 | ||
AK028408.1 | - | 3 - 4 | 441 | 150 | ||
AK028458.1 | - | 3 - 4 | 420 | 150 | ||
AK129053.1 | - | 3 - 4 | 541 | 150 | ||
AK167523.1 | - | 3 - 4 | 477 | 150 | ||
AY225093.1 | - | 3 - 4 | 385 | 150 | ||
AY251192.1 | - | 3 - 4 | 537 | 150 | ||
BC025588.1 | - | 2 - 3 | 372 | 150 | ||
BC030353.1 | - | 3 - 4 | 863 | 150 | ||
BC056968.1 | - | 3 - 4 | 838 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |