Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026665.4 | NP_080941.3 | 9 - 10 | 1332 | 142 | |
NR_037622.1 | 9 - 10 | 1592 | 142 | |||
GenBank mRNA | AK014862.1 | - | 6 - 7 | 965 | 142 | |
AK014873.1 | - | 9 - 10 | 1279 | 142 | ||
AK014982.1 | - | 7 - 8 | 790 | 142 | ||
AK016484.1 | - | 9 - 10 | 1116 | 142 | ||
AK019414.1 | - | 9 - 10 | 1212 | 142 | ||
AK028408.1 | - | 8 - 9 | 1104 | 142 | ||
AY225093.1 | - | 9 - 10 | 1125 | 142 | ||
AY251192.1 | - | 9 - 10 | 1277 | 142 | ||
BC025588.1 | - | 8 - 9 | 1112 | 142 | ||
BC050785.1 | - | 7 - 8 | 843 | 6703205 | 142 | |
BC056968.1 | - | 9 - 10 | 1576 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |