Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmg20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163165.1 | NP_001156637.1 | 4 - 5 | 521 | 69 | |
NM_001163166.1 | NP_001156638.1 | 4 - 5 | 418 | 69 | ||
NM_010440.3 | NP_034570.1 | 3 - 4 | 605 | 69 | ||
GenBank mRNA | AF146224.1 | - | 3 - 4 | 588 | 69 | |
AK153816.1 | - | 4 - 5 | 419 | 69 | ||
AK155615.1 | - | 4 - 5 | 522 | 69 | ||
AK155652.1 | - | 4 - 5 | 423 | 69 | ||
AK155890.1 | - | 3 - 4 | 481 | 69 | ||
AL355734.1 | - | 3 - 4 | 600 | 69 | ||
BC003924.1 | - | 3 - 4 | 493 | 3485302 | 69 | |
BC011334.1 | - | 4 - 5 | 454 | 4012515 | 69 | |
BC071217.1 | - | 1 - 2 | 1482 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |