Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134255.3 | NP_599016.2 | 1 - 2 | 159 | 105 | |
GenBank mRNA | AK028761.1 | - | 1 - 2 | 99 | 105 | |
AK031743.1 | - | 1 - 2 | 161 | 105 | ||
AK045274.1 | - | 1 - 2 | 138 | 105 | ||
AK051580.1 | - | 1 - 2 | 156 | 105 | ||
AK085663.1 | - | 1 - 2 | 122 | 105 | ||
AK085696.1 | - | 1 - 2 | 109 | 105 | ||
AK088153.1 | - | 1 - 2 | 97 | 105 | ||
AK162451.1 | - | 1 - 2 | 160 | 105 | ||
AK167755.1 | - | 1 - 2 | 99 | 105 | ||
AK168360.1 | - | 1 - 2 | 99 | 105 | ||
AK168987.1 | - | 1 - 2 | 86 | 105 | ||
AK170709.1 | - | 1 - 2 | 91 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons |