Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009532.4 | NP_033558.3 | 12 - 13 | 1585 | 65 | |
GenBank mRNA | AK155376.1 | - | 12 - 13 | 1562 | 65 | |
AK165569.1 | - | 12 - 13 | 1540 | 65 | ||
AK168334.1 | - | 12 - 13 | 1535 | 65 | ||
AK168727.1 | - | 12 - 13 | 1539 | 65 | ||
BC055900.1 | - | 12 - 13 | 1561 | 4207343 | 65 | |
BC085281.1 | - | 14 - 15 | 1967 | 30472386 | 65 | |
U02887.1 | - | 12 - 13 | 1549 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |