Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009532.4 | NP_033558.3 | 8 - 9 | 987 | 58 | |
GenBank mRNA | AK155376.1 | - | 8 - 9 | 964 | 58 | |
AK165569.1 | - | 8 - 9 | 942 | 58 | ||
AK168334.1 | - | 8 - 9 | 937 | 58 | ||
AK168727.1 | - | 8 - 9 | 941 | 58 | ||
AK217480.1 | - | 2 - 3 | 230 | 58 | ||
BC055900.1 | - | 8 - 9 | 963 | 4207343 | 58 | |
BC085281.1 | - | 10 - 11 | 1369 | 30472386 | 58 | |
U02887.1 | - | 8 - 9 | 951 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |