Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000018451.mm.1 |
NM_001285435.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000018451.mm.1 |
NR_104339.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000018451.mm.1 |
NR_104340.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000018451.mm.1 |
TC1000002497.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000018457.mm.1 |
NM_001285435.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000018457.mm.1 |
NR_104339.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000018457.mm.1 |
NR_104340.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000018457.mm.1 |
TC1000002497.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000018455.mm.1 |
NM_001285435.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000018455.mm.1 |
NR_104339.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000018455.mm.1 |
NR_104340.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000018455.mm.1 |
TC1000002497.mm.1 |
MTA TC |