Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083912.1 | NP_001077381.1 | 10 - 11 | 1991 | 59 | |
NM_001290542.1 | NP_001277471.1 | 10 - 11 | 2214 | 59 | ||
NM_138752.2 | NP_620091.2 | 10 - 11 | 1994 | 59 | ||
GenBank mRNA | AF465238.1 | - | 10 - 11 | 2459 | 59 | |
AK131110.1 | - | 10 - 11 | 1823 | 59 | ||
AK144152.1 | - | 10 - 11 | 2154 | 59 | ||
BC040403.1 | - | 8 - 9 | 990 | 59 | ||
BC052436.1 | - | 9 - 10 | 1328 | 6837328 | 59 | |
BC056971.1 | - | 10 - 11 | 1262 | 6821362 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |