Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083912.1 | NP_001077381.1 | 6 - 7 | 1411 | 136 | |
NM_001290542.1 | NP_001277471.1 | 6 - 7 | 1634 | 136 | ||
NM_138752.2 | NP_620091.2 | 6 - 7 | 1414 | 136 | ||
GenBank mRNA | AK144152.1 | - | 6 - 7 | 1574 | 136 | |
BC052436.1 | - | 5 - 6 | 748 | 6837328 | 136 | |
BC056971.1 | - | 6 - 7 | 682 | 6821362 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |