Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291915.1 | NP_001278844.1 | 1 - 2 | 126 | 99 | |
GenBank mRNA | BC145412.1 | - | 1 - 2 | 126 | 9053970 | 99 |
DV051476.1 | - | 1 - 2 | 74 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |