Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009163.3 | NP_033189.2 | 11 - 12 | 1251 | 78 | |
GenBank mRNA | AF036894.1 | - | 10 - 11 | 1121 | 78 | |
AK036747.1 | - | 11 - 12 | 1139 | 78 | ||
AK037789.1 | - | 11 - 12 | 1136 | 78 | ||
AK043024.1 | - | 11 - 12 | 1138 | 78 | ||
AK049342.1 | - | 11 - 12 | 1159 | 78 | ||
AK075851.1 | - | 2 - 3 | 236 | 78 | ||
AK150053.1 | - | 11 - 12 | 1146 | 78 | ||
AK173137.1 | - | 11 - 12 | 1180 | 78 | ||
BC026135.1 | - | 11 - 12 | 1197 | 4212293 | 78 | |
CT010214.1 | - | 10 - 11 | 1061 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |