Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183152.3 | NP_898975.2 | 6 - 7 | 757 | 68 | |
GenBank mRNA | AK031968.1 | - | 6 - 7 | 758 | 68 | |
BC086684.1 | - | 1 - 2 | 54 | 30679265 | 68 | |
BC099679.1 | - | 6 - 7 | 726 | 6489647 | 68 | |
BE654673.1 | - | 2 - 3 | 140 | 68 | ||
BG297060.1 | - | 4 - 5 | 344 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |