Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aifm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039194.3 | NP_001034283.1 | 7 - 8 | 881 | 86 | |
NM_001284300.1 | NP_001271229.1 | 7 - 8 | 881 | 86 | ||
NM_153779.2 | NP_722474.2 | 7 - 8 | 874 | 86 | ||
NM_178058.4 | NP_835159.1 | 7 - 8 | 881 | 86 | ||
GenBank mRNA | AK017403.1 | - | 7 - 8 | 878 | 86 | |
AK085656.1 | - | 7 - 8 | 833 | 86 | ||
AK147741.1 | - | 7 - 8 | 858 | 86 | ||
AK155240.1 | - | 7 - 8 | 821 | 86 | ||
BC038129.1 | - | 7 - 8 | 874 | 1395873 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |