Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tirap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177845.1 | NP_001171316.1 | 4 - 5 | 412 | 73 | |
NM_001177846.1 | NP_001171317.1 | 4 - 5 | 394 | 73 | ||
NM_001177847.1 | NP_001171318.1 | 4 - 5 | 340 | 73 | ||
NM_054096.2 | NP_473437.2 | 4 - 5 | 404 | 73 | ||
GenBank mRNA | AF378130.1 | - | 4 - 5 | 313 | 73 | |
AF410784.1 | - | 3 - 4 | 236 | 73 | ||
AK036431.1 | - | 4 - 5 | 333 | 73 | ||
AK080135.1 | - | 4 - 5 | 413 | 73 | ||
AK151117.1 | - | 4 - 5 | 342 | 73 | ||
AK168494.1 | - | 4 - 5 | 332 | 73 | ||
BC005584.1 | - | 1 - 2 | 70 | 3157913 | 73 | |
BY740695.1 | - | 4 - 5 | 400 | 73 | ||
BY743490.1 | - | 4 - 5 | 340 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |