Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038710.1 | NP_001033799.1 | 1 - 2 | 365 | 82 | |
GenBank mRNA | AK015263.1 | - | 1 - 2 | 346 | 82 | |
AK015719.1 | - | 1 - 2 | 401 | 82 | ||
AK017072.1 | - | 1 - 2 | 346 | 82 | ||
AK076562.1 | - | 1 - 2 | 315 | 82 | ||
BC052227.1 | - | 1 - 2 | 365 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |