Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148941.2 | NP_683743.2 | 3 - 4 | 636 | 104 | |
GenBank mRNA | AK029065.1 | - | 3 - 4 | 497 | 104 | |
AK030666.1 | - | 3 - 4 | 585 | 104 | ||
BC037030.1 | - | 3 - 4 | 579 | 4503493 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |