Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301230.1 | NP_001288159.1 | 5 - 6 | 718 | 77 | |
NM_001301232.1 | NP_001288161.1 | 4 - 5 | 611 | 77 | ||
NM_023635.6 | NP_076124.1 | 4 - 5 | 569 | 77 | ||
GenBank mRNA | AB046693.1 | - | 3 - 4 | 367 | 77 | |
AB120511.1 | - | 3 - 4 | 345 | 77 | ||
AB232622.1 | - | 3 - 4 | 345 | 77 | ||
AF304376.1 | - | 4 - 5 | 524 | 77 | ||
AK010373.1 | - | 4 - 5 | 487 | 77 | ||
AK030529.1 | - | 4 - 5 | 516 | 77 | ||
BC008173.1 | - | 5 - 6 | 675 | 3710213 | 77 | |
BC009656.1 | - | 4 - 5 | 522 | 3590089 | 77 | |
CV784419.1 | - | 4 - 5 | 480 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |