Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_028401.1 | 7 - 8 | 1019 | 132 | ||
GenBank mRNA | AK012656.1 | - | 5 - 6 | 868 | 132 | |
AK014920.1 | - | 7 - 8 | 971 | 132 | ||
AK020036.1 | - | 6 - 7 | 931 | 132 | ||
AK050165.1 | - | 7 - 8 | 975 | 132 | ||
AK166329.1 | - | 7 - 8 | 974 | 132 | ||
AK166633.1 | - | 6 - 7 | 2089 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |