Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900001381.mm.1 |
NM_001039520.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001381.mm.1 |
NM_001039520.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001381.mm.1 |
NM_001253893.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001381.mm.1 |
NM_001253893.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001381.mm.1 |
NM_001253894.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001381.mm.1 |
NM_001253894.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001381.mm.1 |
NM_007871.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001381.mm.1 |
NM_007871.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001381.mm.1 |
TC0900000197.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900001376.mm.1 |
NM_001039520.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001376.mm.1 |
NM_001039520.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001376.mm.1 |
NM_001253893.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001376.mm.1 |
NM_001253893.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001376.mm.1 |
NM_001253894.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001376.mm.1 |
NM_001253894.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001376.mm.1 |
NM_007871.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001376.mm.1 |
NM_007871.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001376.mm.1 |
TC0900000197.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900001377.mm.1 |
NM_001039520.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001377.mm.1 |
NM_001039520.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001377.mm.1 |
NM_001253893.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001377.mm.1 |
NM_001253893.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001377.mm.1 |
NM_001253894.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001377.mm.1 |
NM_001253894.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001377.mm.1 |
NM_007871.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001377.mm.1 |
NM_007871.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001377.mm.1 |
TC0900000197.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900001383.mm.1 |
NM_001039520.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001383.mm.1 |
NM_001039520.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001383.mm.1 |
NM_001253893.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001383.mm.1 |
NM_001253893.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001383.mm.1 |
NM_001253894.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001383.mm.1 |
NM_001253894.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001383.mm.1 |
NM_007871.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001383.mm.1 |
NM_007871.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900001383.mm.1 |
TC0900000197.mm.1 |
MTA TC |