Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293650.1 | NP_001280579.1 | 13 - 14 | 1336 | 71 | |
NM_001293651.1 | NP_001280580.1 | 13 - 14 | 1319 | 71 | ||
NM_008925.2 | NP_032951.1 | 13 - 14 | 1315 | 71 | ||
GenBank mRNA | AK137661.1 | - | 13 - 14 | 1282 | 71 | |
AK153931.1 | - | 13 - 14 | 1290 | 71 | ||
AK177012.1 | - | 2 - 3 | 106 | 71 | ||
AK208791.1 | - | 1 - 2 | 56 | 71 | ||
BC009816.1 | - | 13 - 14 | 1269 | 4008182 | 71 | |
CX207653.1 | - | 5 - 6 | 455 | 71 | ||
U92794.1 | - | 13 - 14 | 1268 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |