Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Rgl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023622.4 | NP_076111.2 | 1 - 2 | 87 | 75 | |
GenBank mRNA | AF237669.1 | - | 1 - 2 | 46 | 75 | |
AF239661.1 | - | 1 - 2 | 88 | 75 | ||
AK004876.1 | - | 1 - 2 | 50 | 75 | ||
AK053550.1 | - | 1 - 2 | 73 | 75 | ||
AK134654.1 | - | 69 | 75 | |||
AK165023.1 | - | 1 - 2 | 80 | 75 | ||
AK165746.1 | - | 1 - 2 | 80 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus 1700 Probe spans exons |