Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023622.4 | NP_076111.2 | 4 - 5 | 484 | 70 | |
GenBank mRNA | AF237669.1 | - | 4 - 5 | 443 | 70 | |
AF239661.1 | - | 4 - 5 | 485 | 70 | ||
AK004876.1 | - | 3 - 4 | 217 | 70 | ||
AK053550.1 | - | 4 - 5 | 471 | 70 | ||
AK134654.1 | - | 466 | 70 | |||
AK165023.1 | - | 4 - 5 | 472 | 70 | ||
AK165746.1 | - | 4 - 5 | 477 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |