Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ulk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027895.1 | NP_082171.1 | 12 - 13 | 1376 | 101 | |
GenBank mRNA | AK004783.1 | - | 10 - 11 | 1958 | 101 | |
AK032030.1 | - | 10 - 11 | 998 | 101 | ||
AK154642.1 | - | 12 - 13 | 1289 | 101 | ||
BC039958.1 | - | 8 - 9 | 738 | 101 | ||
BC151153.1 | - | 12 - 13 | 1302 | 9088047 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |