Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ulk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027895.1 | NP_082171.1 | 7 - 8 | 983 | 87 | |
GenBank mRNA | AK004783.1 | - | 5 - 6 | 1565 | 87 | |
AK032030.1 | - | 5 - 6 | 605 | 87 | ||
AK154642.1 | - | 7 - 8 | 896 | 87 | ||
AK186089.1 | - | 3 - 4 | 88 | |||
BC037093.1 | - | 7 - 8 | 1004 | 87 | ||
BC039958.1 | - | 3 - 4 | 345 | 87 | ||
BC151153.1 | - | 7 - 8 | 909 | 9088047 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |