Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ulk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027895.1 | NP_082171.1 | 11 - 12 | 1305 | 78 | |
GenBank mRNA | AK004783.1 | - | 9 - 10 | 1887 | 78 | |
AK032030.1 | - | 9 - 10 | 927 | 78 | ||
AK154642.1 | - | 11 - 12 | 1218 | 78 | ||
BC037093.1 | - | 11 - 12 | 1326 | 78 | ||
BC039958.1 | - | 7 - 8 | 667 | 78 | ||
BC151153.1 | - | 11 - 12 | 1231 | 9088047 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |