Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900004265.mm.1 |
NM_028300.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004265.mm.1 |
NM_028300.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004265.mm.1 |
TC0900000602.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900004266.mm.1 |
NM_028300.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004266.mm.1 |
NM_028300.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004266.mm.1 |
TC0900000602.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900004268.mm.1 |
NM_028300.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004268.mm.1 |
NM_028300.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004268.mm.1 |
TC0900000602.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900004264.mm.1 |
NM_028300.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004264.mm.1 |
NM_028300.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004264.mm.1 |
TC0900000602.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900004274.mm.1 |
NM_028300.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004274.mm.1 |
NM_028300.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004274.mm.1 |
TC0900000602.mm.1 |
MTA TC |