Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dixdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178118.2 | NP_835219.1 | 13 - 14 | 1580 | 67 | |
GenBank mRNA | AK142668.1 | - | 7 - 8 | 704 | 67 | |
AK147565.1 | - | 7 - 8 | 2320 | 67 | ||
AK147596.1 | - | 7 - 8 | 2320 | 67 | ||
AY549883.1 | - | 13 - 14 | 1438 | 67 | ||
AY549884.1 | - | 13 - 14 | 1441 | 67 | ||
AY549885.1 | - | 9 - 10 | 730 | 67 | ||
AY549886.1 | - | 9 - 10 | 739 | 67 | ||
AY549887.1 | - | 7 - 8 | 481 | 67 | ||
BC048182.1 | - | 13 - 14 | 1571 | 6826299 | 67 | |
BC063085.1 | - | 13 - 14 | 1571 | 6826299 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |