Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081346.1 | NP_001074815.1 | 4 - 5 | 531 | 76 | |
GenBank mRNA | AK030986.1 | - | 4 - 5 | 415 | 76 | |
AK045134.1 | - | 432 | 76 | |||
AK052662.1 | - | 4 - 5 | 569 | 76 | ||
BC116341.2 | - | 533 | 40098130 | 76 | ||
BC116342.2 | - | 533 | 40098131 | 76 | ||
BC158093.1 | - | 4 - 5 | 536 | 9007380 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |