Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK030986.1 | - | 6 - 7 | 720 | 63 | |
AK045134.1 | - | 737 | 63 | |||
AK052662.1 | - | 6 - 7 | 874 | 63 | ||
BC116341.2 | - | 838 | 40098130 | 63 | ||
BC116342.2 | - | 838 | 40098131 | 63 | ||
BC158093.1 | - | 6 - 7 | 841 | 9007380 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |