Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153059.1 | NP_694699.1 | 4 - 5 | 777 | 65 | |
GenBank mRNA | AK043098.1 | - | 811 | 65 | ||
AK075928.1 | - | 4 - 5 | 767 | 65 | ||
AK078061.1 | - | 4 - 5 | 845 | 65 | ||
BC020100.1 | - | 4 - 5 | 777 | 3981114 | 65 | |
BC023194.1 | - | 1 - 2 | 225 | 65 | ||
BC083125.1 | - | 5 - 6 | 945 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |