Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029364.3 | NP_083640.1 | 6 - 7 | 873 | 69 | |
GenBank mRNA | AK030773.1 | - | 6 - 7 | 872 | 69 | |
AK049162.1 | - | 6 - 7 | 874 | 69 | ||
AK054046.1 | - | 6 - 7 | 873 | 69 | ||
AK083597.1 | - | 6 - 7 | 851 | 69 | ||
AK149863.1 | - | 6 - 7 | 858 | 69 | ||
AK151037.1 | - | 4 - 5 | 495 | 69 | ||
AK159562.1 | - | 6 - 7 | 840 | 69 | ||
AK165180.1 | - | 6 - 7 | 853 | 69 | ||
AK165917.1 | - | 6 - 7 | 823 | 69 | ||
AK169485.1 | - | 6 - 7 | 855 | 69 | ||
AK170791.1 | - | 6 - 7 | 869 | 69 | ||
AK181505.1 | - | 1 - 2 | 72 | 69 | ||
BC055328.1 | - | 6 - 7 | 867 | 6400020 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |