Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lemd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081193.2 | NP_001074662.2 | 6 - 7 | 1948 | 96 | |
GenBank mRNA | AK140538.1 | - | 6 - 7 | 431 | 96 | |
BC082610.1 | - | 7 - 8 | 712 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |