Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029057.1 | NP_083333.1 | 2 - 3 | 608 | 68 | |
GenBank mRNA | AK044521.1 | - | 2 - 3 | 415 | 68 | |
AK049989.1 | - | 2 - 3 | 440 | 68 | ||
AK173079.1 | - | 2 - 3 | 371 | 68 | ||
BC072607.1 | - | 2 - 3 | 435 | 68 | ||
BC138285.1 | - | 2 - 3 | 815 | 8861304 | 68 | |
BC138286.1 | - | 2 - 3 | 815 | 8861305 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |