Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P197402 |
BC059081.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P197402 |
BC072607.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P197402 |
NM_029057.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P197402 |
BC059081.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P197402 |
BC072607.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P197402 |
NM_029057.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
325118 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
8785 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
970753 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
104942 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
833033 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
676690 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
201001 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
524627 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
160466 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776213 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
236233 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
36849 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807626 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
813592 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454305 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
582339 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
343272 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
45780 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
698678 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
359935 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
524303 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
285272 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
678553 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
562154 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
493117 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
446333 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
836336 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
135343 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
246064 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
310249 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
6965 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829258 |
AK080736.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829258 |
AK129163.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829258 |
AK143802.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829258 |
AK149826.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829258 |
BC046331.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829258 |
BC060106.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829258 |
NM_177673.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
151494 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
70293 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
116859 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
126587 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
77842 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
45661 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P118494 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P105112 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P197402 |
BC059081.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P197402 |
BC072607.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P197402 |
NM_029057.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P197402 |
BC059081.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P197402 |
BC072607.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P197402 |
NM_029057.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02235341 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02235396 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P118494 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P105112 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P197402 |
BC059081.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P197402 |
BC072607.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P197402 |
NM_029057.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P197402 |
BC059081.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P197402 |
BC072607.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P197402 |
NM_029057.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262616 |
BC138286.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262616 |
BC138285.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262616 |
AK173079.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262616 |
BC059081.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262616 |
BC072607.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P105112 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P118494 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502447 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08575303 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502435 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502762 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502722 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502678 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502750 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502439 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502655 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502785 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502547 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08431328 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08575290 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502826 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08445796 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08248010 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502810 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502822 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502653 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502727 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08731782 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502806 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08881997 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502699 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502672 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502530 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08873900 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502745 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502834 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502829 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502733 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502707 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08224011 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502747 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502830 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08633549 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502796 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502676 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502648 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502528 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502813 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08248005 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502697 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502548 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08432481 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08560019 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502544 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502797 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502554 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502709 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502835 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502685 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502758 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502551 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502817 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502433 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08328725 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08575295 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502809 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502708 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502431 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08779112 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502764 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08968832 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502558 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502775 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502791 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502801 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502799 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08445797 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502519 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08565278 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502650 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502824 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502735 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502781 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502694 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502702 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502545 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502782 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502533 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502442 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502737 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502842 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502743 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502712 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502754 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502746 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502684 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502444 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502773 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502450 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502770 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502769 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08477287 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502837 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502833 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08532608 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502808 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502729 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502449 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502667 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08575300 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502744 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502790 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502748 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08543242 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502441 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502553 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502434 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08543243 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502752 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502644 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502717 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502713 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502724 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502696 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502803 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502683 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502823 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502776 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08669905 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08224014 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08511163 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08511160 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08731787 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08575297 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502559 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08809361 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502682 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08445793 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502759 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502841 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502756 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502706 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502704 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502542 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502840 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08232138 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08575306 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502774 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502664 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502814 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502688 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502668 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502642 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502657 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08328722 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502720 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502777 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502787 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08224018 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502780 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08565272 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502784 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502549 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502715 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502662 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502660 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502673 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502818 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502793 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502757 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502445 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08857226 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27502451 |
- |
- |