Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wee2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201370.2 | NP_958758.2 | 1 - 2 | 317 | 67 | |
GenBank mRNA | AK139573.1 | - | 1 - 2 | 276 | 67 | |
AK143336.1 | - | 1 - 2 | 241 | 67 | ||
AK163344.1 | - | 1 - 2 | 319 | 67 | ||
BC080784.1 | - | 1 - 2 | 242 | 30481521 | 67 | |
BY725757.1 | - | 1 - 2 | 318 | 67 | ||
DQ011691.1 | - | 1 - 2 | 138 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |