Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK029800.1 | - | 1 - 2 | 119 | 78 | |
AK086574.1 | - | 1 - 2 | 119 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |