Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163085.2 | NP_001156557.2 | 20 - 21 | 2781 | 86 | |
GenBank mRNA | BC031147.1 | - | 19 - 20 | 2286 | 86 | |
BC043943.1 | - | 19 - 20 | 2286 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |