Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081056.1 | NP_001074525.1 | 17 - 18 | 2265 | 77 | |
GenBank mRNA | AK029113.1 | - | 12 - 13 | 1717 | 77 | |
BC094337.1 | - | 2 - 3 | 284 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |